Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: EXPLOSIVE 1.4G

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

Manufacturer:  Brady™ 63366

 View more versions of this product

Catalog No. 19-111-294


Add to cart

Description

Description

  • Available in two different materials Premium Fiberglass, Pressure Sensitive Vinyl
Specifications

Specifications

Non-adhesive
EXPLOSIVE 1.4G
Diamond
0.0984 in
Vehicle Placard
Black, Orange
Material Handling, Shipping & Receiving Signs & Labels
W x H: 10.75 x 10.75 in.
Fiberglass
1/ea
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit