Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: (Flammable Picto w/Blank Box) 3

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

Specifications

Size (English) W x H: 10.75 x 10.75 in.
Product Type Vehicle Placard
Display Text (Flammable picto w/ blank box) 3
For Use With (Application) Material Handling, Shipping and Receiving Signs and Labels
Color Red, White
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19-110-998
View Documents
Brady™
60379
Pressure Sensitive Vinyl (B-946) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-111-166
View Documents
Brady™
60411
Premium Fiberglass (B-120) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Available in two different materials Pressure Sensitive Vinyl and Premium Fiberglass
Specifications

Specifications

W x H: 10.75 x 10.75 in.
(Flammable picto w/ blank box) 3
Red, White
Vehicle Placard
Material Handling, Shipping and Receiving Signs and Labels
Diamond
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit