Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ DOT Vehicle Placards: SPONTANEOUSLY COMBUSTIBLE 4
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
Specifications
Adhesive Type | Non-adhesive |
---|---|
Color | Black, Red, White |
Product Type | Vehicle Placard |
Display Text | SPONTANEOUSLY COMBUSTIBLE 4 |
For Use With (Application) | Material Handling, Shipping and Receiving Signs and Labels |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
19-111-335
|
Brady™
63426 |
Fiberglass (B-120) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-111-350
|
Brady™
63447 |
Polycoated Tagstock (B-101) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Available in two different materials Polycoated Tagstock and Premium Fiberglass
Specifications
Non-adhesive | |
Vehicle Placard | |
Material Handling, Shipping and Receiving Signs and Labels | |
Diamond |
Black, Red, White | |
SPONTANEOUSLY COMBUSTIBLE 4 | |
1 Ea. | |
10.75 x 10.75 in. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title