Learn More
Eisco™ Double Concave Glass Lens
Perform experiments with high quality, optically true lenses. The Double Concave Glass Lens is made of optically worked, highly polished glass with ground edges.
Supplier: Eisco™ PH0536RCC
Description
- Spherical
- Optically worked glass
- Ground edges
- Highly polished
Specifications
Glass | |
3 in. | |
300 mm | |
Focal length: 500 mm |
75 mm | |
For physics classroom demonstrations in optics and light refraction | |
Double Concave Lens |