Learn More
Sklar InstrumentsSupplier Diversity Partner Double Ended Probe, Sklar, 7 in., Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 301570
Double Ended Probe, Sklar, 7 in., Stainless Steel. Sklar's Double Ended Probe is an instrument used to assist in retrieving information such as depth or size. The probe is used to determine weather foreign bodies or other issues exist. The probe is available in a variety of lengths and tip sizes.
Catalog No. 50-143-8137