Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Double stranded DNA Mouse anti-Human, Clone: AE-2, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14632
Description
Mouse monoclonal antibody raised against human double stranded DNA.
- High specificity
- High reactivity
Specifications
Double stranded DNA | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunocytochemistry (0.5-1 ug/mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determin | |
Mouse | |
Protein G purification | |
RUO | |
This antibody recognizes the double stranded DNA in human cells. | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG3 κ |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
AE-2 | |
Mouse monoclonal antibody raised against human double stranded DNA. | |
In 10 mM PBS. | |
Nuclei of Burkitt's cells. | |
100 μg | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction