Learn More
Doublecortin Antibody (OTI4A3), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270558UV
Catalog No. NBP270558UV
Add to Cart
Description
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Doublecortin
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
DCX
Full length recombinant protein of human Doublecortin (NP_835365) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI4A3
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin)
Mouse
Affinity Purified
Neuroscience, Phospho Specific, Signal Transduction
1641.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions