Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Doublecortin Monoclonal Antibody (01), Invitrogen™
Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | Doublecortin |
---|---|
Clone | 01 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543489
|
Invitrogen™
MA543489 |
50 μL |
Each for $285.50
|
|
|||||
PIMA543039
|
Invitrogen™
MA543039 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human Doublecortin/DCX
In the developing cortex, cortical neurons must migrate over long distances to reach the site of their final differentiation. DCX is a cytoplasmic protein which appears to direct neuronal migration by regulating the organization and stability of microtubules. It contains two doublecortin domains, which bind microtubules. In addition, this protein interacts with LIS1, the regulatory gamma subunit of platelet activating factor acetylhydrolase, and this interaction is important to proper microtubule function in the developing cortex. Mutations in the gene encoding DCX are a cause of X-linked lissencephaly.Specifications
Doublecortin | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
DBCN; Dbct; DC; Dcn; DCX; doublecortex; doublecortin; doublin; lissencephalin-X; lissencephaly, X-linked (doublecortin); LISX; Lis-X; LIS-XD; Neuronal migration protein doublecortin; OTTHUMP00000023860; OTTHUMP00000216316; SCLH; XLIS | |
DCX | |
IgG2a | |
Protein A | |
Antibody |
01 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
O43602 | |
1641 | |
Recombinant Human Doublecortin/DCX Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
DCX |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title