Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doublecortin Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI4A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270558JF549

Catalog No. N270558J549


Add to cart

Description

Description

Doublecortin Monoclonal antibody specifically detects Doublecortin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Doublecortin
OTI4A3
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
DCX
Full length recombinant protein of human Doublecortin (NP_835365) produced in HEK293T cells.
Immunogen affinity purified
Neuroscience, Phospho Specific, Signal Transduction
Primary
1641.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin)
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit