Learn More
Selleck Chemical LLCSupplier Diversity Partner Dovitinib (TKI-258) 25mg 405169-16-6 CHIR-258  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S101825MG
Dovitinib (TKI258, CHIR258) is a multitargeted RTK inhibitor, mostly for class III (FLT3/c-Kit) with IC50 of 1 nM/2 nM, also potent to class IV (FGFR1/3) and class V (VEGFR1-4) RTKs with IC50 of 8-13 nM, less potent to InsR, EGFR, c-Met, EphA2, Tie2, IGF-1R and HER2 in cell-free assays. Phase 4.
Catalog No. 50-619-7