Learn More
Dow Corning™ Pharma-50 Tubing
Translucent, pharmaceutical-grade silicone tubing for ultrapure fluid processing.
Manufacturer:  DOW CORNING CORPORATION 3100511
 View more versions of this product
Catalog No. 14-177-113
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Platinum Cured Silicone
1 in.
0.125 in.
25.4mm
For ultrapure liquid transfer
3.2mm
Yes
0.75 in.
19.1mm
Exceeds USP Class VI Plastic Test requirements and meets EP monograph “Silicone Elastomer for Closures and Tubing”. Manufactured according to Bulk Pharmaceutical Chemicals GMPs at an FDA-registered, ISO 9002-certified healthcare facility.
50 ft.