Learn More
Dow Corning™ Silastic™ Laboratory Tubing
Flexible, translucent silicone elastomer tubing
Manufacturer:  DOW CORNING CORPORATION 2415682
 View more versions of this product
Catalog No. 11-189-13C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ISO 9001, FDA, USP Class VI
0.38 in.
9.7mm
Fluid processing, separation and filtration such as drug manufacturing, oxygen delivery in production processes and chemical, pharmaceutical and environmental analyses
15.3m
-54° to 249°C
2.38mm
Yes
Silicone
Translucent
0.56 in.
14.2mm
Peristaltic pump
-65.2° to 480°F
0.094 in.
2.38mm (0.094 in.)
50 ft.