Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dow Corning™ Silastic™ Laboratory Tubing

Flexible, translucent silicone elastomer tubing

Manufacturer:  DOW CORNING CORPORATION 2415607

 View more versions of this product

Catalog No. 11-189-15L


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ISO 9001, FDA, USP Class VI, 3A Sanitary standard
0.13 in.
3.3mm
Fluid processing, separation and filtration such as drug manufacturing, oxygen delivery in production processes and chemical, pharmaceutical and environmental analyses
15.3m
-54° to 249°C
1.57mm
Yes
Silicone
Translucent
0.25 in.
6.35mm
Peristaltic pump
-65.2° to 480°F
0.062 in.
1.57mm (0.0625 in.)
50 ft.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit