Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 1X2-200, ion-exchange resin, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
9085-42-1
C29H34ClN
432.048
MFCD00132715
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl,dowex 1x8 50-100 cl
16212807
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
9085-42-1
C29H34ClN
432.048
MFCD00132715
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl,dowex 1x8 50-100 cl
16212807
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
Particle Size Particle size analysis: on 50 mesh (cumulative): < 15%, through 140 mesh: < 15%
Water Volume Capacity wet volume capacity, Cl form (meq/mL):> 0.6
Water Retention Capacity Cl form: 70 to 80%
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Merck Index 10, 2182
Solubility Information Solubility in water: insoluble.
Quantity 2.5kg
Physical Form Beads
Chemical Name or Material Dowex™ 1X2