Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 1X2, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
9085-42-1
C29H34ClN
432.048
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
16212807
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
9085-42-1
C29H34ClN
432.048
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
16212807
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
Ion exchange resin
Additional Information Particle size analysis: on 35 mesh (cumulative): 10% max., through 80 mesh: 10% max
Packaging Plastic bottle
Water Retention Capacity 65 to 75% (Cl form)
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Quantity 500g
Solubility Information Solubility in water: insoluble.
Physical Form Beads
Chemical Name or Material Dowex™ 1X2