Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 1X8-100, ion-exchange resin, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
12627-85-9
C29H34ClN
432.048
MFCD00132718
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl,dowex 1x8 50-100 cl
16212807
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
12627-85-9
C29H34ClN
432.048
MFCD00132718
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl,dowex 1x8 50-100 cl
16212807
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]

Dowex 1X8, C29H34ClN, CAS Number-12627-85-9, dowex 1x8 50-100 cl, dowex 1x2 50-100 cl, 500g, White, 01,511; 04,267; 06,302; 07,182; 08,263, BBQMUEOYPPPODD-UHFFFAOYSA-M, 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride, MFCD00132718, 432.048, Plastic bottle

Dowex 1X8, C29H34ClN, CAS Number-12627-85-9, dowex 1x8 50-100 cl, dowex 1x2 50-100 cl, 500g, White, 01,511; 04,267; 06,302; 07,182; 08,263, BBQMUEOYPPPODD-UHFFFAOYSA-M, 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride, MFCD00132718, 432.048, Plastic bottle

Ion exchange resin
Particle Size 15 % max. (on 35 mesh, cumulative) 15 % max. (through 80 mesh)
Packaging Plastic bottle
Water Volume Capacity wet volume capacity, Cl form (meq/mL):> 1.2
Water Retention Capacity Cl form: 43 to 48%
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Color White
Quantity 500g
Physical Form Powder and/or Chunks
Chemical Name or Material Dowex™ 1X8