Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 50WX8-100 ion-exchange resin, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic Bottle
11119-67-8
MFCD00132726
11119-67-8
MFCD00132726
Particle Size 15% max. (on 35 mesh, cumulative), 20% max. (through 80 mesh)
Packaging Plastic Bottle
Water Volume Capacity wet volume capacity, H form min. (meq/mL): 1.7
Water Retention Capacity H form: 50 to 56%
Fieser 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256
Color Beige to Tan or Green-Tan
Solubility Information Solubility in water: insoluble.
Quantity 500g
Physical Form Moist Spheres
Chemical Name or Material Dowex™ 50WX8