Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 50WX8-400 ion-exchange resin, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic Bottle
11119-67-8
MFCD00132726
11119-67-8
MFCD00132726
Particle Size 15% max. (on 100 mesh, cumulative), 15% max. (through 325 mesh)
Packaging Plastic Bottle
Water Volume Capacity wet volume capacity, H form min. (meq/mL): 1.7
Water Retention Capacity H form: 50 to 58%
Color Beige to Brown
Solubility Information Solubility in water: insoluble.
Quantity 500g
Physical Form Beads
Chemical Name or Material Dowex™ 50WX8