Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 5WX2-200, ion-exchange resin, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic Bottle
12612-37-2
MFCD00132722
12612-37-2
MFCD00132722
Particle Size Particle size analysis: on 35 mesh (cumulative): 15% max., through 140 mesh: 15% max.
Packaging Plastic Bottle
Water Volume Capacity wet volume capacity, H form min. (meq/mL): 0.6
Water Retention Capacity H form: 74 to 82%
Quantity 100g
Physical Form Beads
Chemical Name or Material Dowex™ 50WX2