Learn More
Mystaire™ Downflow Workstation Main Filter Available on GSA/VA Contract for Federal Government customers only.
For use with the Mystaire™ Downflow Workstation (FE-WS6). Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
Manufacturer:  Mystaire™ FE5004
 View more versions of this product
Catalog No. 15-338-192
Add to cart
Description
Description
  • Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
For Hydrocarbons- FE-WS6, Hydrocarbons
Carbon filter
FE-WS6
Gas Phase Carbon