Learn More
Mystaire™ Downflow Workstation Main Filter
For use with the Mystaire™ Downflow Workstation (FE-WS6). Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
Manufacturer:  Mystaire™ FE5014
 View more versions of this product
Catalog No. 15-338-194
Add to cart
Description
Description
  • Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
Specifications
For Mineral Acids- FE-WS6, Mineral Acids
Carbon filter
FE-WS6
Gas Phase Carbon