Learn More
Medchemexpress LLC Doxazosin mesylate | 77883-43-3 | 98.5% | C24H29N5O8S | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0098A100MG
Encompass_Preferred
Doxazosin mesylate is a quinazoline-derivative that selectively antagonizes postsynaptic α1-adrenergic receptors. It is the mesylate salt form of doxazosin, a long-lasting inhibitor of α1-adrenoceptors. This product is for research use only and is not sold to patients.
  • Selectively antagonizes postsynaptic α1-adrenergic receptors
  • Long-lasting inhibitor of α1-adrenoceptors
  • Used to treat benign prostatic hyperplasia and lower urinary tract symptoms
  • May have a direct inhibitory effect on cholesterol synthesis
Catalog No. 50-004-32715