Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxepin Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
1229-29-4
C19H22ClNO
315.84
MHNSPTUQQIYJOT-SJDTYFKWSA-N
hydrogen dimethyl({3-[(2E)-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene]propyl})amine chloride
[H+].[Cl-].CN(C)CC\C=C1/C2=CC=CC=C2COC2=CC=CC=C12
1229-29-4
C19H22ClNO
315.84
MHNSPTUQQIYJOT-SJDTYFKWSA-N
hydrogen dimethyl({3-[(2E)-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene]propyl})amine chloride
[H+].[Cl-].CN(C)CC\C=C1/C2=CC=CC=C2COC2=CC=CC=C12

D1195, 1229-29-4, C19H21NO•HCl

Spectrum™ Doxepin Hydrochloride, USP is a tricyclic antidepressant used to treat mental and mood problems such as depression and anxiety. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.002
Loss on Drying 0.005
Quantity 100 g
CAS Max % 1
UN Number UN2811
Percent Purity 98 to 102%