Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Doxorubicin, Hydrochloride, ≥98%, Calbiochem™,

Click to view available options
Quantity:
10mg
Packaging:
Glass Bottle
25316-40-9
C27H30ClNO11
579.98
MFCD00077757,MFCD00077757,MFCD00941448
MWWSFMDVAYGXBV-FGBJBKNOSA-N
doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
129626538
hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
[H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
25316-40-9
C27H30ClNO11
579.98
MFCD00077757,MFCD00077757,MFCD00941448
MWWSFMDVAYGXBV-FGBJBKNOSA-N
doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
129626538
hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
[H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Recommended Storage

15°C to 30°C, Stock solutions are Stable for up to 1 month at 4°C. Under acidic conditions doxorubicin is converted to a water−insoluble adriamycinone and a water soluble reducing sugar, daunosamine.

Color Red
Packaging Glass Bottle
Quantity 10mg
Percent Purity ≥98% by HPLC

Recommended Storage : 15°C to 30°C, Stock solutions are Stable for up to 1 month at 4°C. Under acidic conditions doxorubicin is converted to a water−insoluble adriamycinone and a water soluble reducing sugar, daunosamine.