Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxorubicin Hydrochloride, Fisher BioReagents

Click to view available options
Quantity:
1mg
5mg
10mg
50mg
Packaging:
Amber Glass
25316-40-9
C27H30ClNO11
579.98
MFCD00077757,MFCD00077757,MFCD00941448
MWWSFMDVAYGXBV-FGBJBKNOSA-N
doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
129626538
(7R,9R)-7-[(2S,4R,5R,6R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
[H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
25316-40-9
C27H30ClNO11
579.98
MFCD00077757,MFCD00077757,MFCD00941448
MWWSFMDVAYGXBV-FGBJBKNOSA-N
doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
129626538
(7R,9R)-7-[(2S,4R,5R,6R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
[H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Doxorubicin Hydrochloride

Doxorubicin HCL is a chemotherapeutic agent. Antitumor, immunosuppressive, and antibiotic agent. It blocks RNA polymerase and reverse transcriptase. Also inhibits nucleic acid synthesis.
Color Orange
Melting Point 216°C
Quantity 1mg
Decomposition Information 216°C
Packaging Amber Glass
Water 4.0% max.
Assay Percent Range (by HPLC): ≥95%
Merck Index 15, 3487
Solubility Information Soluble in water
Formula Weight 580
Physical Form Powder/Solid
Chemical Name or Material Doxorubicin Hydrochloride
Show More Show Less

WARNING!

Emergency Overview
May cause cancer. Risk of serious damage to eyes. Irritating to respiratory system and skin. May cause heritable genetic damage. Possible risk of impaired fertility. Possible risk of harm to the unborn child. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:1
Instability:0

Recommended Storage : 2° to 8°C