Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Doxycycline hyclate

Click to view available options
Quantity:
25g
100g
24390-14-5
C26H41ClN2O12
609.066
MFCD07357237
ISZOFWGVNMFENH-YDLUHMIOSA-N
Hydramycin; Doxycycline hydrochloride hemiethanolate hemihydrate
131676157
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride
CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl
24390-14-5
C26H41ClN2O12
609.066
MFCD07357237
ISZOFWGVNMFENH-YDLUHMIOSA-N
Hydramycin; Doxycycline hydrochloride hemiethanolate hemihydrate
131676157
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride
CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl

Inhibits matrix metalloproteinases at subantimicrobial doses

Applications
Inhibits matrix metalloproteinases at subantimicrobial doses.Doxycycline hyclate is an antibacterial antibiotic, which is used in the treatment of chlamydia, rickettsia, mycoplasma and some spirochete infections. It is also used to inhibit matrix metalloproteinases at subantimicrobial doses.

Solubility
Soluble in water to 50mg/ml. May require warming.Soluble in water, methanol. Sparingly soluble in ethanol. Insoluble in chloroform and ether.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Merck Index 14,3440
Solubility Information Soluble in water to 50mg/ml; May require warming
Formula Weight 512.94 (503.94 Anhydrous)
Physical Form Powder
Color Yellow
Quantity 25g
Chemical Name or Material Doxycycline hyclate

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QI8925000

Recommended Storage : 2° to 8°C