Learn More
Reprocell Stemolecule Doxycycline hyclate, 24390-14-5, MFCD07357237, 10 mg
Supplier: Reprocell 040016
Empirical Formula (Hill Notation): C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O, Molecular Weight: 512.94, Synonym: Doxycycline hydrochloride hemiethanolate hemihydrate. Yellow to yellow with a green cast powder, store powder at 4 °C protected from light.