Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxycycline Hyclate, USP, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

D2993, 24390-14-5, (C22H24N2O8•HCl)2•C2H6O•H2O

Spectrum™ Doxycycline Hyclate, USP is tetracycline antibiotic used to treat a wide variety of bacterial infections. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Quantity 25 kg
CAS 24390-14-5
CAS Max % 1
Molecular Formula C46H58Cl2N4O18
InChI Key QDDHZFKCHWYXJF-PCLMCNLLSA-N
SMILES [H+].[H+].O.[Cl-].[Cl-].CCO.C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H](N(C)C)C(=O)\C(=C(\N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12.C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H](N(C)C)C(=O)\C(=C(\N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12
IUPAC Name dihydrogen bis((2E,4S,4aR,5S,5aR,6R,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-5,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione) ethanol hydrate dichloride
Molecular Weight (g/mol) 1025.88