Learn More
Medchemexpress LLC Doxycycline hydrochloride | 10592-13-9 | MFCD03427564 | 100.0% | 480.90 | C22H25ClN2O8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0565AR10MG
Encompass_Preferred
Doxycycline hydrochloride is the hydrochloride salt of doxycycline used in research as an antibiotic and broad-spectrum metalloproteinase (MMP) inhibitor. It is used in antibacterial assays, anti-proliferative studies, and tetracycline-regulated gene expression systems.
  • Research-grade antibiotic and MMP inhibitor.
  • Exhibits antibacterial and anti-cancer cell proliferation activity.
  • Suitable for constructing inducible gene expression models.
  • High purity for analytical and biological applications.
  • Molecular formula C22H25ClN2O8; molecular weight 480.90.
Catalog No. 50-002-96834