Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxycycline hydrochloride, 20 mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
50mL
100mL
10592-13-9
C22H25ClN2O8
480.898
MFCD03427564
PTNZGHXUZDHMIQ-CVHRZJFOSA-N
54706018
[(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)[NH+](C)C)O.[Cl-]
10592-13-9
C22H25ClN2O8
480.898
MFCD03427564
PTNZGHXUZDHMIQ-CVHRZJFOSA-N
54706018
[(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)[NH+](C)C)O.[Cl-]
Packaging Plastic bottle
Physical Form Liquid
Quantity 100mL
Chemical Name or Material Doxycycline hydrochloride