Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxylamine succinate, 98%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
562-10-7
C21H28N2O5
388.464
KBAUFVUYFNWQFM-UHFFFAOYSA-N
doxylamine succinate,decapryn,evigoa d,a-h injection,decapryn succinate,unisom,alsadorm,gittalun,mereprine
11224
CHEBI:82461
butanedioic acid;N,N-dimethyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine
CC(C1=CC=CC=C1)(C2=CC=CC=N2)OCCN(C)C.C(CC(=O)O)C(=O)O
562-10-7
C21H28N2O5
388.464
KBAUFVUYFNWQFM-UHFFFAOYSA-N
doxylamine succinate,decapryn,evigoa d,a-h injection,decapryn succinate,unisom,alsadorm,gittalun,mereprine
11224
CHEBI:82461
butanedioic acid;N,N-dimethyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine
CC(C1=CC=CC=C1)(C2=CC=CC=N2)OCCN(C)C.C(CC(=O)O)C(=O)O
Packaging Glass bottle
Quantity 5g
Assay Percent Range 98%
Formula Weight 388.46
Chemical Name or Material Doxylamine succinate