Learn More
Sklar InstrumentsSupplier Diversity Partner Doyen Rib Raspatory, Sklar, 7 in., Blunt, Angled (Left), Smooth, Child / Pediatric / Small, Hollow Octogen, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 227760
Doyen Rib Raspatory, Sklar, 7 in., Blunt, Angled (Left), Smooth, Child / Pediatric / Small, Hollow Octogen, Stainless Steel. Sklar's Doyen Raspatory has a spiraled hook shape to scrape individual ribs. The handle has a hollow grip for better control. The instrument is available angled left or right as well as adult and pediatric sizes used during cardiothoracic surgery procedures.
Catalog No. 50-143-7878