Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ DP-1 Transcription Factor Ab-6, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting experiments with Thermo Scientific™ DP-1 Transcription Factor Ab-6, Mouse Monoclonal Antibody.

$190.00 - $935.00

Specifications

Antigen DP-1 Transcription Factor Ab-6
Clone 1DP06; same as TFD10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
MS1056P
View Documents
Thermo Scientific™
MS1056P
200μg/mL 1mL Each for $885.50
Add to cart
 
MS1056P0
View Documents
Thermo Scientific™
MS1056P0
200μg/mL 100μL Each for $190.00
Add to cart
 
MS1056P1
View Documents
Thermo Scientific™
MS1056P1
200μg/mL 500μL Each for $523.50
Add to cart
 
MS1056P1ABX
View Documents
Thermo Scientific™
MS1056P1ABX
1mg/mL 100μL Each for $597.50
Add to cart
 
MS1056PABX
View Documents
Thermo Scientific™
MS1056PABX
1mg/mL 200μL Each for $935.00
Add to cart
 
MS1056R1
View Documents
Thermo Scientific™
MS1056R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in western blotting experiments with Thermo Scientific™ DP-1 Transcription Factor Ab-6, Mouse Monoclonal Antibody.

DP family contains at least two members, DP-1 and -2. Structurally, DP-1 and -2 are relatively conserved, yet their expression appears to be tissue specific. E2F-1, a functional target for the tumor suppressor protein Rb, heterodimers with DP-1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.

Host Species: Mouse

Clone: 1DP06 (TFD10)

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 83-204

Immunogen: Recombinant DP-1 protein

Molecular Weight: 45 kDa

Positive Control: MAD109 or Raji cells, Tonsil

Cellular Localization: Cytoplasm and nucleus

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

DP-1 Transcription Factor Ab-6
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Murine
Transcription Regulation
Recombinant DP-1 protein
1DP06; same as TFD10
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit