Learn More
Supply SolutionsSupplier Diversity Partner Hach - 10ml DPD Free Chlorine Reagent SwifTest; Platform: Swiftest; Parameter: Chlorine, Free; Range: 0.02 to 2.00 mgL Cl2; Number of tests: 250; Method: 8021; EPA compliant: Yes DFS Item
Manufacturer:  Supply SolutionsSupplier Diversity Partner HACH 2105560
For free chlorine determination by the DPD Free Chlorine method. For use with Hach Swiftest DPD dispenser. USEPA accepted for reporting for drinking water analysis using Hach Method 8021. Total chlorine and potassium iodide refill vials also available. Range: 0.02 - 2.00 mgL Cl2. Sample size: 10ml. Approximately 250 tests per vial. This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.; Platform: Swiftest; Parameter: Chlorine, Free; Range: 0.02 to 2.00 mgL Cl2; Number of tests: 250; Method: 8021; EPA compliant: Yes; Method Name: DPD; Just squeeze to add powder
Catalog No. 50-476-336