Learn More
Supply SolutionsSupplier Diversity Partner Hach Dpd Free Chlorine Reagent Accuvac Ampules. For Use With: Determination Of Free Chlorine.; Grade: Reagent; Concentration: 0.02-2Mg/L Cl2 DFS Item
Manufacturer:  Supply SolutionsSupplier Diversity Partner HACH 2502025
For determination of Free Chlorine by the DPD Method, using AccuVac Ampules. USEPA accepted for reporting drinking water analysis using Hach Method 8021. Range: 0.02 - 2.00 mgL Cl2. Package of 25 ampules. This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.; Grade: Reagent; Concentration: 0.02-2mg/L Cl2; For use with: Determination of Free Chlorine; Platform : Accuvac; Method Name: DPD; Range: 0.02 to 2.00 mgL Cl2; Number of tests: 25; Method: 8021; Instrument: All; EPA compliant: Yes; Fast
Catalog No. 50-476-301