Learn More
DPD Indicator Solution, for Residual Chlorine Analysis, Ricca Chemical
For Analysis of Residual Chlorine Analysis
Supplier: Ricca Chemical Company 26551
Specifications
DPD Indicator Solution | |
100.0, 0.34, 0.1, 0.02 | |
Amber Glass Bottle |
98.0, 0.33, 0.1, 0.02 | |
7664-93-9, 6283-63-2, 6381-92-6 | |
4 L |
Safety and Handling
Recommended Storage : Refrigerate (2°C - 8°C)