DPD Indicator Solution, for Residual Chlorine Analysis, Ricca Chemical

For Analysis of Residual Chlorine Analysis

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 265516

Packaging Amber Glass
Quantity 500mL

 View more versions of this product

Catalog No. 2655-16

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,7664-93-9,6283-63-2,6381-92-6
    CAS Min % 98.0,0.33,0.1,0.02
    CAS Max % 100.0,0.34,0.1,0.02
    Chemical Name or Material DPD Indicator Solution

    Specifications

    Quantity 500mL
    CAS 6381-92-6,6283-63-2,7732-18-5,7664-93-9
    CAS Max % 0.02,0.34,0.1,100.0
    CAS Min % 98.0,0.33,0.02,0.1
    Recommended Storage Refrigerate (2°C - 8°C)
    Packaging Amber Glass
    Chemical Name or Material DPD Indicator Solution

    Refrigerate (2°C - 8°C)

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit