Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPEP2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549481
Description
Probable metalloprotease which hydrolyzes leukotriene D4 (LTD4) into leukotriene E4 (LTE4).Specifications
DPEP2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9H4A9 | |
DPEP2 | |
KLH conjugated synthetic peptide between 138-171 amino acids from human DPEP2. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DPEP2 | |
Dipeptidase 2, DPEP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64174 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction