Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPEP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 164661AP150UL
Description
Probable metalloprotease which hydrolyzes leukotriene D4 (LTD4) into leukotriene E4 (LTE4).Specifications
DPEP2 | |
Polyclonal | |
Unconjugated | |
DPEP2 | |
dipeptidase 2, DPEP2, MBD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291984, 319446, 64174 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5M872, Q8C255, Q9H4A9 | |
Dpep2 | |
DPEP2 Fusion Protein Ag9545 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction