Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPH2 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI3A12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270587AF700

Catalog No. N270587A700


Add to cart

Description

Description

DPH2 Monoclonal antibody specifically detects DPH2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

DPH2
OTI3A12
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
DPH2
Full length recombinant protein of human DPH2 (NP_001375) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
diphthamide biosynthesis protein 2, Diphthamide biosynthesis protein 2 homolog-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2(S. cerevisiae), DPH2 homolog, DPH2 homolog (S. cerevisiae), DPH2L2diphthamide biosynthesis-like protein 2, DPH2-like 2, DPH2-like 2 (S. cerevisiae), HsDph2
Mouse
IgG1
0.1 mL
Primary
1802.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit