Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPH2 Mouse anti-Human, Mouse, Rat, Canine, Monkey, PE, Clone: OTI4E7, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270585PE
Description
DPH2 Monoclonal antibody specifically detects DPH2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.Specifications
DPH2 | |
OTI4E7 | |
Flow Cytometry | |
Affinity Purified, Conjugated | |
DPH2 | |
Full length recombinant protein of human DPH2 (NP_001375) produced in HEK293T cells. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Canine, Human, Monkey, Mouse, Rat |
Flow Cytometry | |
PE | |
PBS with 0.05% Sodium Azide | |
diphthamide biosynthesis protein 2, Diphthamide biosynthesis protein 2 homolog-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2(S. cerevisiae), DPH2 homolog, DPH2 homolog (S. cerevisiae), DPH2L2diphthamide biosynthesis-like protein 2, DPH2-like 2, DPH2-like 2 (S. cerevisiae), HsDph2 | |
Mouse | |
IgG1 | |
0.1 mL | |
Primary | |
1802.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title