Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DPM1 Monoclonal Antibody (5C5A7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ A6429

Description
DPM1 Monoclonal Antibody for Western Blot
Dolichol-phosphate mannose (Dol-P-Man) serves as a donor of mannosyl residues on the lumenal side of the endoplasmic reticulum (ER). Lack of Dol-P-Man results in defective surface expression of GPI-anchored proteins. Dol-P-Man is synthesized from GDP-mannose and dolichol-phosphate on the cytosolic side of the ER by the enzyme dolichyl-phosphate mannosyltransferase. Human DPM1 lacks a carboxy-terminal transmembrane domain and signal sequence and is regulated by DPM2.
Specifications
DPM1 | |
Monoclonal | |
1 mg/mL | |
HEPES buffered saline with 0.02% sodium azide | |
P14020 | |
DPM1 | |
Yeast Dolichol phosphate mannose synthase. | |
250 μg | |
Primary | |
Yeast | |
Antibody | |
IgG1 |
Western Blot | |
5C5A7 | |
Unconjugated | |
DPM1 | |
AI118379; AI194292; CDGIE; dolichol monophosphate mannose synthase; dolichol-phosphate (beta-D) mannosyltransferase 1; Dolichol-phosphate mannose synthase; Dolichol-phosphate mannose synthase subunit 1; Dolichol-phosphate mannosyltransferase; Dolichol-phosphate mannosyltransferase subunit 1; dolichyl-phosphate beta-D-mannosyltransferase; Dolichyl-phosphate beta-D-mannosyltransferase subunit 1; dolichyl-phosphate mannosyltransferase 1; dolichyl-phosphate mannosyltransferase polypeptide 1 catalytic subunit; dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit; DPM synthase; DPM synthase complex, catalytic subunit; DPM synthase subunit 1; Dpm1; Mannose-P-dolichol synthase; Mannose-P-dolichol synthase subunit 1; MPD synthase; MPD synthase subunit 1; MPDS; OTTMUSP00000017164; P9705.3; RP23-391M18.1; SED3; YPR183W | |
Mouse | |
Purified | |
RUO | |
856313 | |
4°C | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction