Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPP10 Antibody (OTI1A5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272089H
Description
DPP10 Monoclonal specifically detects DPP10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DPP10 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
DPP10 | |
Full length recombinant protein of human DPP10 (NP_065919) produced in HEK293T cells. | |
0.1 mL | |
57628.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1A5 | |
Western Blot, Immunohistochemistry | |
dipeptidyl peptidase 10, Dipeptidyl peptidase IV-related protein 3, dipeptidyl peptidase like protein 2, Dipeptidyl peptidase X, Dipeptidyl peptidase-like protein 2, dipeptidyl-peptidase 10, dipeptidyl-peptidase 10 (non-functional), DPL2, DPP X, DPPY, DPRP-3, DPRP3dipeptidylpeptidase 10, inactive dipeptidyl peptidase 10, KIAA1492 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction