Learn More
DPP2 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS6045R
Catalog No. 50-198-7001
Add to cart
Description
Description
The protein encoded by this gene is a post-proline cleaving aminopeptidase expressed in quiescent lymphocytes. The resting lymphocytes are maintained through suppression of apoptosis, a state which is disrupted by inhibition of this novel serine protease. The enzyme has strong sequence homology with prolylcarboxypeptidase and is active at both acidic and neutral pH.
Specifications
Specifications
DPP2
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DPP7
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
29952
DPP7
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q9UHL4
carboxytripeptidase; Dipeptidyl aminopeptidase II; dipeptidyl arylamidase II; Dipeptidyl peptidase 2; Dipeptidyl peptidase 7; Dipeptidyl peptidase II; dipeptidyl-peptidase 2; dipeptidylpeptidase 7; dipeptidyl-peptidase II; DPP II; Dpp2; Dpp7; DPPII; QPP; Quiescent cell proline dipeptidase
KLH conjugated synthetic peptide derived from human DPP2.
Protein A
RUO
Antibody
Polyclonal
Human