Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP3 Mouse anti-Human, PE/Cy5.5, Clone: OTI2D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271541PECY55

Catalog No. N271541PEC5


Add to cart

Description

Description

DPP3 Monoclonal antibody specifically detects DPP3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

DPP3
OTI2D9
Flow Cytometry
Affinity Purified, Conjugated
DPP3
Full length recombinant protein of human DPP3 (NP_005691) produced in HEK293 cell.
Immunogen affinity purified
Core ESC Like Genes, Stem Cell Markers
Primary
10072.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
Dipeptidyl aminopeptidase III, Dipeptidyl arylamidase III, dipeptidyl peptidase 3, Dipeptidyl peptidase III, dipeptidylpeptidase 3, dipeptidyl-peptidase 3, dipeptidylpeptidase III, DPP III, DPPIII, EC 3.4.14.4, FLJ11387, FLJ22331
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit