Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP3 Mouse anti-Human, Clone: OTI 2G8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525638

Catalog No. PIMA525638


Add to cart

Description

Description

This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternatively spliced transcript variants have been found for this gene.
Specifications

Specifications

DPP3
Monoclonal
0.13 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NY33
Dipeptidyl peptidase 3; Dipeptidyl aminopeptidase III; Dipeptidyl arylamidase III; Dipeptidyl peptidase III; DPP III; Enkephalinase B; DPP3
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10072
Flow Cytometry, Western Blot
OTI 2G8
Unconjugated
DPP3
Liquid
DPP3
Full length human recombit protein of DPP3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit