Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP9 Mouse anti-Human, Clone: OTI2D4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503959

Catalog No. 50-167-4733


Add to cart

Description

Description

This gene encodes a protein that is a member of the S9B family in clan SC of the serine proteases. The protein has been shown to have post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Although the activity of this protein is similar to that of dipeptidyl peptidase 4 (DPP4), it does not appear to be membrane bound. In general, dipeptidyl peptidases appear to be involved in the regulation of the activity of their substrates and have been linked to a variety of diseases including type 2 diabetes, obesity and cancer. Several transcript variants of this gene have been described but not fully characterized.
Specifications

Specifications

DPP9
Monoclonal
0.68 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q86TI2
DP9, DPLP9, DPRP-2, DPRP2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
91039
Immunocytochemistry, Immunofluorescence, Western Blot
OTI2D4
Unconjugated
DPP9
Liquid
DPP9
Full length human recombit protein of human DPP9 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit