Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPA2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15463120UL

 View more versions of this product

Catalog No. NBP1546320


Add to cart

Description

Description

DPPA2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

DPPA2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
DPPA2
Synthetic peptides corresponding to DPPA2(developmental pluripotency associated 2) The peptide sequence was selected from the N terminal of DPPA2. Peptide sequence NMEQMEPSVSSTSDVKLEKPKKYNPGHLLQTNEQFTAPQKARCKIPALPL.
34 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:100-1:2000
Q7Z7J5
CT100, developmental pluripotency associated 2, developmental pluripotency-associated protein 2, PESCRG1cancer/testis antigen 100, Pluripotent embryonic stem cell-related gene 1 protein
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
151871
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit