Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPE fumarate, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

1185241-83-1
C23H29NO5
399.487
ZOOMDYHTGZSGDD-WLHGVMLRSA-N
N, N-Diethyl-2-[4-(phenylmethyl)phenoxy]ethanamine fumarate
45073473
2-(4-benzylphenoxy)-N,N-diethylethanamine;(E)-but-2-enedioic acid
CCN(CC)CCOC1=CC=C(C=C1)CC2=CC=CC=C2.C(=CC(=O)O)C(=O)O

Inhibitor of histamine binding at the intracellular binding site

Binds with high affinity to the antiestrogen binding site but not to the estrogen receptor.?Induces of sterol accumulation in MCF7 cells. An inhibitor of histamine binding at the intracellular histamine (Hlc) site.
Quantity 50mg
Formula Weight 399.49
Chemical Name or Material DPPE fumarate
Grade >98%

Recommended Storage : Desiccate at -20°C