Learn More
DPPIV/CD26 Antibody (OTI11D7), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270588IR
Catalog No. NBP270588IR
Add to Cart
Description
Description
DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
DPPIV/CD26
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
DPP4
Full length recombinant protein of human DPPIV/CD26 (NP_001926) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI11D7
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5
Mouse
Affinity Purified
Adaptive Immunity, Cellular Markers, GPCR, Immunology
1803.0
Store at 4°C in the dark.
Recommendations
Recommendations